Learn More
Optimize Technologies OPTI-SOLV™ In-Line Replacement Frits, 2.0µm, 0.125”Diameter, 10pk
SDP
Catalog No. 502006156
Encompass_Preferred
The user-friendly OPTI-SOLV™ In-Line Filter system is well suited for a number of applications, such as high temperature, high-pressure environments where quick changes are imperative. The unique swivel design allows frits to be changed without removing the connecting tubing. Just plumb it once and leave it. The frictional forces generated at the piston/seal interface inevitably lead to particulate shedding. Placement of a filtering element between the pump and injector/ auto sampler removes these particulates, preventing problems with downstream components and costly downtime. Key Features include 10 Filter Elements, a 316 Stainless steel frit (solvent compatible), and an FEP sealing surface. Use with OPTI-SOLV In-Line Filter Holders and the Agilent Purge Valve Upgrade Kit :: Optimize Part numbers: 10-04-03685 & 10-04-03686.
Catalog No. 50-200-6156 Supplier Optimize Technologies Supplier No. 100403702
The user-friendly OPTI-SOLV™ In-Line Filter system is well suited for a number of applications, such as high temperature, high-pressure environments where quick changes are imperative. The unique swivel design allows frits to be changed without removing the connecting tubing. Just plumb it once and leave it. The frictional forces generated at the piston/seal interface inevitably lead to particulate shedding. Placement of a filtering element between the pump and injector/ auto sampler removes these particulates, preventing problems with downstream components and costly downtime. Key Features include 10 Filter Elements, a 316 Stainless steel frit (solvent compatible), and an FEP sealing surface. Use with OPTI-SOLV In-Line Filter Holders and the Agilent Purge Valve Upgrade Kit :: Optimize Part numbers: 10-04-03685 & 10-04-03686.