Learn More
LUKNOVA INC Porous Polyethylene Frits For Luknova Supersep 330 Ml/240 Gram Empty Flash Column. It Has Pore Size 20 µm And Disc Diametr Ø 65 Mm. Each Unit Comes With 50 Top Ffrits And 50 Bottom Frits.
Catalog No. 50920685
Encompass
Porous Polyethylene Frits For Luknova Supersep 330 Ml/240 Gram Empty Flash Column. It Has Pore Size 20 µm And Disc Diametr Ø 65 Mm. Each Unit Comes With 50 Top Ffrits And 50 Bottom Frits.
Catalog No. 50-920-685 Supplier LUKNOVA INC Supplier No. FC0032407
Porous Polyethylene Frits For Luknova Supersep 330 Ml/240 Gram Empty Flash Column. It Has Pore Size 20 µm And Disc Diametr Ø 65 Mm. Each Unit Comes With 50 Top Ffrits And 50 Bottom Frits.