Learn More
Thermo Scientific Copy IconThermo Scientific™UltiMate™ 3000 Pump Filter Frit
Catalog No. 501348074 Shop All Thermo Scientific Products
Encompass
Stainless steel, prevent contaminant particles from reaching your HPLC system. Thermo Scientific™ Dionex™ UltiMate™ 3000 Pump Filter Frits remove contaminants that may be present in your solvent supply and prevent them from fouling your LC system. These frits are available in three different materials to match your chromatography needs.
Catalog No. 50-134-8074 Supplier Thermo Scientific Supplier No. 6268.0110
Stainless steel, prevent contaminant particles from reaching your HPLC system. Thermo Scientific™ Dionex™ UltiMate™ 3000 Pump Filter Frits remove contaminants that may be present in your solvent supply and prevent them from fouling your LC system. These frits are available in three different materials to match your chromatography needs.