Learn More
Chemglass Life Sciences Fritted Disc, 40mm, Coarse, Filter
SDP
Catalog No. 50873559
Encompass_Preferred
Fritted Disc, 40mm, Coarse, Filter. Porous discs for use in fabrication of special glass apparatus. Available in F, M, C and XC porosities.Pressure Limits:Fritted glassware is designed for low pressure vacuum filtration or gas flow. The MAX differential pressure should not exceed 15 PSI.Thermal Limits:Fritted glassware should not be exposed to excessive temperature change or direct exposure to flame due to the fact that it has less resistance to thermal shock than standard borosilicate glassware.
Catalog No. 50-873-559 Supplier Chemglass Life Sciences Supplier No. CG20131
Fritted Disc, 40mm, Coarse, Filter. Porous discs for use in fabrication of special glass apparatus. Available in F, M, C and XC porosities.Pressure Limits:Fritted glassware is designed for low pressure vacuum filtration or gas flow. The MAX differential pressure should not exceed 15 PSI.Thermal Limits:Fritted glassware should not be exposed to excessive temperature change or direct exposure to flame due to the fact that it has less resistance to thermal shock than standard borosilicate glassware.