Learn More
Chemglass Life Sciences Fritted Disc, 10mm, Extra Coarse, Filter
SDP
Catalog No. CG20104
Encompass_Preferred
Fritted Disc, 10mm, Extra Coarse Porosity, Filter. Porous discs for use in fabrication of special glass apparatus.Pressure Limits:Fritted glassware is designed for low pressure vacuum filtration or gas flow. The MAX differential pressure should not exceed 15 PSI.Thermal Limits:Fritted glassware should not be exposed to excessive temperature change or direct exposure to flame due to the fact that it has less resistance to thermal shock than standard borosilicate glassware.
Catalog No. CG20104 Supplier Chemglass Life Sciences Supplier No. CG20104
Fritted Disc, 10mm, Extra Coarse Porosity, Filter. Porous discs for use in fabrication of special glass apparatus.Pressure Limits:Fritted glassware is designed for low pressure vacuum filtration or gas flow. The MAX differential pressure should not exceed 15 PSI.Thermal Limits:Fritted glassware should not be exposed to excessive temperature change or direct exposure to flame due to the fact that it has less resistance to thermal shock than standard borosilicate glassware.