Learn More
Medchemexpress LLC Frizzled-10/CD350 M 100ug
SDP
Catalog No. 5000179951
Encompass_Preferred
Frizzled-10/CD350 protein is a receptor for Wnt proteins that functions in the canonical Wnt/ -catenin pathway activating disheveled proteins inhibiting GSK-3 kinase and triggering Wnt target gene activation Secondary signaling pathways involve PKC and calcium flux Frizzled-10/CD350 Protein Mouse (HEK293 His) is the recombinant mouse-derived Frizzled-10/CD350 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79951 Supplier Medchemexpress LLC Supplier No. HYP74144100UG
Frizzled-10/CD350 protein is a receptor for Wnt proteins that functions in the canonical Wnt/ -catenin pathway activating disheveled proteins inhibiting GSK-3 kinase and triggering Wnt target gene activation Secondary signaling pathways involve PKC and calcium flux Frizzled-10/CD350 Protein Mouse (HEK293 His) is the recombinant mouse-derived Frizzled-10/CD350 protein expressed by HEK293 with C-His labeled tag