Learn More
Medchemexpress LLC Frizzled-10/CD350 M 10ug
SDP
Catalog No. 5000179098
Encompass_Preferred
Frizzled-10/CD350 protein is a receptor for Wnt proteins that functions in the canonical Wnt/ -catenin pathway activating disheveled proteins inhibiting GSK-3 kinase and triggering Wnt target gene activation Secondary signaling pathways involve PKC and calcium flux Frizzled-10/CD350 Protein Mouse (HEK293 His) is the recombinant mouse-derived Frizzled-10/CD350 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79098 Supplier Medchemexpress LLC Supplier No. HYP7414410UG
Frizzled-10/CD350 protein is a receptor for Wnt proteins that functions in the canonical Wnt/ -catenin pathway activating disheveled proteins inhibiting GSK-3 kinase and triggering Wnt target gene activation Secondary signaling pathways involve PKC and calcium flux Frizzled-10/CD350 Protein Mouse (HEK293 His) is the recombinant mouse-derived Frizzled-10/CD350 protein expressed by HEK293 with C-His labeled tag