Learn More
ACROBiosystems ACROBiosystems Human Frizzled-2 / FZD2 Protein, Fc Tag
Catalog No. 501957121
Encompass_Preferred
Recombinant Protein;100UG;Human Frizzled-2, Fc Tag (FZ2-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Ser 156 (Accession # AAI13403).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.7 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7121 Supplier ACROBiosystems Supplier No. FZ2H5251100UG
Recombinant Protein;100UG;Human Frizzled-2, Fc Tag (FZ2-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Ser 156 (Accession # AAI13403).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.7 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.