Learn More
ACROBiosystems ACROBiosystems Human Frizzled-2 / FZD2 (24-190) Protein, Fc Tag
Catalog No. 501957201
Encompass_Preferred
Recombinant Protein;1MG;Human Frizzled-2, Fc Tag (FZ2-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Pro 190 (Accession # Q14332-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.4 kDa. The protein migrates as 50-62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7201 Supplier ACROBiosystems Supplier No. FZ2H52551MG
Recombinant Protein;1MG;Human Frizzled-2, Fc Tag (FZ2-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Pro 190 (Accession # Q14332-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.4 kDa. The protein migrates as 50-62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.