Learn More
Medchemexpress LLC Frizzled-4/CD344 Hu 50ug
SDP
Catalog No. 5000179903
Encompass_Preferred
The Frizzled-4/CD344 protein is a receptor for Wnt proteins and is associated with the classical -catenin pathway activating disheveled proteins inhibiting GSK-3 kinase and triggering Wnt target genes In retinal vascularization it acts as a receptor for Wnt proteins and Norrin (NDP) stimulating -catenin accumulation and LEF/TCF-mediated transcription Frizzled-4/CD344 Protein Human (HEK293 His) is the recombinant human-derived Frizzled-4/CD344 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79903 Supplier Medchemexpress LLC Supplier No. HYP7414050UG
Add to Cart
Add to Cart
The Frizzled-4/CD344 protein is a receptor for Wnt proteins and is associated with the classical -catenin pathway activating disheveled proteins inhibiting GSK-3 kinase and triggering Wnt target genes In retinal vascularization it acts as a receptor for Wnt proteins and Norrin (NDP) stimulating -catenin accumulation and LEF/TCF-mediated transcription Frizzled-4/CD344 Protein Human (HEK293 His) is the recombinant human-derived Frizzled-4/CD344 protein expressed by HEK293 with C-His labeled tag