Learn More
ACROBiosystems ACROBiosystems Human Frizzled-7 / FZD7 Protein, His Tag
Catalog No. 501956707
Encompass_Preferred
Recombinant Protein;100UG;Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 33 - Leu 185 (Accession # O75084-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 25-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6707 Supplier ACROBiosystems Supplier No. FZ7H52H9100UG
Recombinant Protein;100UG;Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 33 - Leu 185 (Accession # O75084-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 25-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.