Learn More
ACROBiosystems Recombinant Protein;Human Frizzled-4 / FZD4 Protein, Fc Tag;HEK293;100UG;FZ4,FZD4,Frizzled-4,FZD-4,Frizzled 4,CD344,Fz-4,hFz4,FzE4,Frizzled 4
Catalog No. 501658519
Encompass_Preferred
Recombinant Protein;Human Frizzled-4 / FZD4 Protein, Fc Tag;100UG;Human Frizzled-4, Fc Tag (FZ4-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 37 - Glu 180 (Accession # AAI14528).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8519 Supplier ACROBiosystems Supplier No. FZ4H5250100UG
Recombinant Protein;Human Frizzled-4 / FZD4 Protein, Fc Tag;100UG;Human Frizzled-4, Fc Tag (FZ4-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 37 - Glu 180 (Accession # AAI14528).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.