Learn More
Sino Biological Recombinant Human FGFR4 Protein (GST Tag), Active
Supplier:  Sino Biological F0711G500
Encompass_Preferred
Recombinant human FGFR4 (460-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.
Catalog No. 50-257-4900