Learn More
Aviva Systems Biology Corp FRK Antibody - middle region (ARP54618_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54618_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FRK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FRK. Alternate names: GTK, PTK5, RAKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61262