Learn More
Newcomer Supply Formalex Green pH Test Strips, 100 strips/pack
SDP
Catalog No. NC1425229
Encompass
For determining the completion of the reaction of the treated waste formalin with Formalex Green.  Once treated solution is at or above pH 5.5 it can be safely disposed.
Catalog No. NC1425229 Supplier Newcomer Supply Supplier No. ABFXGPH
For determining the completion of the reaction of the treated waste formalin with Formalex Green.  Once treated solution is at or above pH 5.5 it can be safely disposed.