Learn More
Electron Microscopy Sciences Formvar/Carbon Film 3-4 nm/3-4 nm thick on Thin Bar Hexagonal 200 mesh Nickel Grid
SDP
Catalog No. 501926112
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is 5-6 nm thick with a 3 to 4 nm carbon coating.
Grid: Nickel
Catalog No. 50-192-6112 Supplier Electron Microscopy Sciences Supplier No. FCFTH200NIUB
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is 5-6 nm thick with a 3 to 4 nm carbon coating.
Grid: Nickel