Learn More
Electron Microscopy Sciences Formvar/Carbon Film 25-50 nm/20-30 nm thick on Hexagonal 100 mesh Copper Grid
SDP
Catalog No. 501903637
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is 25 to 50 nm thick with a 20 to 30 nm carbon coating.
Grid: Copper
Catalog No. 50-190-3637 Supplier Electron Microscopy Sciences Supplier No. FCF100HCUEC
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is 25 to 50 nm thick with a 20 to 30 nm carbon coating.
Grid: Copper