Learn More
Electron Microscopy Sciences Formvar/Carbon Film 10 nm/20-30 nm thick on Hexagonal 300 mesh Nickel Grid
SDP
Catalog No. 501925912
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is approximately 10 nm thick with 20 to 30 nm of carbon.
Grid: Nickel
Catalog No. 50-192-5912 Supplier Electron Microscopy Sciences Supplier No. FCF300HNISC
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is approximately 10 nm thick with 20 to 30 nm of carbon.
Grid: Nickel