Learn More
Electron Microscopy Sciences Formvar/Carbon Film 10 nm/1 nm thick on Single 200 µm hole Nickel Grid
SDP
Catalog No. 501901046
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is approximately 10 nm thick with 1 nm of carbon.
Grid: Nickel
Catalog No. 50-190-1046 Supplier Electron Microscopy Sciences Supplier No. FCFGA200NI25
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is approximately 10 nm thick with 1 nm of carbon.
Grid: Nickel