Learn More
Electron Microscopy Sciences Formvar/Carbon Support Film 15-20 nm/3-4 nm thick on London Finder LF400 Nickel Grid
SDP
Catalog No. 501903622
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is 15 to 20 nm thick with a 3 to 4 nm carbon coating.
Grid: Nickel
Catalog No. 50-190-3622 Supplier Electron Microscopy Sciences Supplier No. FCFLF400NITB
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is 15 to 20 nm thick with a 3 to 4 nm carbon coating.
Grid: Nickel