Learn More
Electron Microscopy Sciences Formvar/Carbon Film 15-20 nm/3-4 nm thick on Square 150 mesh Gold Grid
SDP
Catalog No. 501903383
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Formvar is 15 to 20 nm thick with a 3 to 4 nm carbon coating.
Grid: Gold
Catalog No. 50-190-3383 Supplier Electron Microscopy Sciences Supplier No. FCF150AUTB
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Formvar is 15 to 20 nm thick with a 3 to 4 nm carbon coating.
Grid: Gold