Learn More
Electron Microscopy Sciences Formvar/Carbon Film 10 nm/3-4 nm thick on Thin Bar Square 200 mesh Gold Grid
SDP
Catalog No. 501925988
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is approximately 10 nm thick with 3 to 4 nm of carbon.
Grid: Gold
Catalog No. 50-192-5988 Supplier Electron Microscopy Sciences Supplier No. FCFT200AUSB
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is approximately 10 nm thick with 3 to 4 nm of carbon.
Grid: Gold