Learn More
Electron Microscopy Sciences Formvar/Carbon Film 25-50 nm/1 nm thick on Square 400 mesh Gold Grid
SDP
Catalog No. 501903481
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Formvar is 25 to 50 nm thick with a 1 nm carbon coating.
Grid: Gold
Catalog No. 50-190-3481 Supplier Electron Microscopy Sciences Supplier No. FCF400AUEA
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Formvar is 25 to 50 nm thick with a 1 nm carbon coating.
Grid: Gold