Learn More
Electron Microscopy Sciences Formvar/Carbon Film 5-6 nm/1 nm thick on Square 300 mesh Copper Grid
SDP
Supplier:  Electron Microscopy Sciences FCF300CUUA
Encompass_Preferred
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid. Formvar is 5-6 nm thick with a 1 nm carbon coating.
Grid: Copper
Catalog No. NC1200385