Learn More
Electron Microscopy Sciences Formvar Support Film 20-30 nm thick on Square 300 mesh Gold Grid
SDP
Catalog No. 501902832
Encompass_Preferred
Support Film on grids has become a main product line for us since the demand for high quality coated grids has increased. To make your microscopy work easier and to save you a great deal of time we offer you a complete line. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold
Catalog No. 50-190-2832 Supplier Electron Microscopy Sciences Supplier No. FF300AUET
Support Film on grids has become a main product line for us since the demand for high quality coated grids has increased. To make your microscopy work easier and to save you a great deal of time we offer you a complete line. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold