Learn More
Electron Microscopy Sciences Formvar Support Film 10 nm thick on London Finder LF135 Copper Grid
SDP
Catalog No. 501902861
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a London finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper
Catalog No. 50-190-2861 Supplier Electron Microscopy Sciences Supplier No. FFLF135CUTH
A thin film of pure formvar resin coats the shiny side of a London finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper