Learn More
Electron Microscopy Sciences Formvar Support Film 10 nm thick on Single 800 µm hole Copper Grid
SDP
Catalog No. 501903050
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper
Catalog No. 50-190-3050 Supplier Electron Microscopy Sciences Supplier No. FFGA800CUTH
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper