Learn More
Electron Microscopy Sciences Formvar Support Film 20-30 nm thick on Single 800 µm hole Copper Grid
SDP
Catalog No. 501903051
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Copper
Catalog No. 50-190-3051 Supplier Electron Microscopy Sciences Supplier No. FFGA800CUET
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Copper