Learn More
Electron Microscopy Sciences Formvar Support Film 3-4 nm thick on Single 150 µm hole Copper Grid
SDP
Catalog No. 501903019
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper
Catalog No. 50-190-3019 Supplier Electron Microscopy Sciences Supplier No. FFGA150CUUL
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper