Learn More
Electron Microscopy Sciences Formvar Support Film 10 nm thick on Thin Bar Hexagonal 300 mesh Copper Grid
SDP
Catalog No. 501902963
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a thin bar hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper
Catalog No. 50-190-2963 Supplier Electron Microscopy Sciences Supplier No. FFTH300CUTH
A thin film of pure formvar resin coats the shiny side of a thin bar hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Copper