Learn More
MSC Cooper Bussmann FRN-R-1 125 VDC, 250 VAC, 1 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 50.8mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 9/16" Diam
Supplier:  MSC 75784678
Encompass
Cooper Bussmann FRN-R-1 125 VDC, 250 VAC, 1 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 50.8mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 9/16" Diam
Catalog No. 50-130-5027