Learn More
MSC Cooper Bussmann FRN-R-60 125 VDC, 250 VAC, 60 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 76.2mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 13/16" Diam
Catalog No. 501305033
Encompass
Cooper Bussmann FRN-R-60 125 VDC, 250 VAC, 60 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 76.2mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 13/16" Diam
Catalog No. 50-130-5033 Supplier MSC Supplier No. 75908350
Cooper Bussmann FRN-R-60 125 VDC, 250 VAC, 60 Amp, Time Delay General Purpose Fuse Fuse Holder Mount, 76.2mm Overall Length, 20 at DC, 200 (RMS) kA Rating, 13/16" Diam