Learn More
Electron Microscopy Sciences Fro-Marker for Frozen Sections
SDP
Catalog No. 50930981
Encompass_Preferred
A wide tipped pen designed for use with frozen sections. The pen provides a sticky membrane which when applied to the slide and onto which the section is placed it eliminates the section from falling off, moving, or wrinkling on the slide during immunostaining procedures. The membrane is stable up to 110 degree C and is suitable for vigorous applications such as in-situ hybridization.
Catalog No. 50-930-981 Supplier Electron Microscopy Sciences Supplier No. 7131310
A wide tipped pen designed for use with frozen sections. The pen provides a sticky membrane which when applied to the slide and onto which the section is placed it eliminates the section from falling off, moving, or wrinkling on the slide during immunostaining procedures. The membrane is stable up to 110 degree C and is suitable for vigorous applications such as in-situ hybridization.