Learn More
Lakeland Industries Lakeland CleanMax Microporous Cleanroom frock. ISO Class 4-6 cleanrooms and all controlled environments. Clean manufactured. Individually packaged. 30 per case with overlay bag. Size 5X
Catalog No. NC2875473
Encompass_Preferred
CleanMax Microporous is available in sterile and non-sterile configurations. Clean-Manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax meets IEST Particle Category Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. Resistant to microbial, viral and blood borne contaminants, along with some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier for the wearer. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of a strong contamination control strategy. Frocks are constructed with microporous film laminate material that is fluid repellant and protects against breakthrough and strikethrough. Mandarin collar, zipper with placket, elastic wrist with thumb loops. Individually packaged, 30/case
Catalog No. NC2875473 Supplier Lakeland Industries Supplier No. CTL191CM5X
CleanMax Microporous is available in sterile and non-sterile configurations. Clean-Manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax meets IEST Particle Category Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. Resistant to microbial, viral and blood borne contaminants, along with some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier for the wearer. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of a strong contamination control strategy. Frocks are constructed with microporous film laminate material that is fluid repellant and protects against breakthrough and strikethrough. Mandarin collar, zipper with placket, elastic wrist with thumb loops. Individually packaged, 30/case