Learn More
3B Scientific™ Frog Embryology Slide Set
SDPSCI_ED
Catalog No. S95066L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
10
Catalog No. Quantity
S95066L 10
1 options
Catalog No. S95066L Supplier 3B Scientific™ Supplier No. W13056
Add to Cart
Add to Cart
Rana
  • Frog, morula, l.s. with macro- and micromeres
  • Frog, blastula. l.s. shows blastocoel
  • Frog, gastrula, sagittal l.s. shows germ layers, dorsal lip, yolk plug
  • Frog, neurula, t.s. showing primordium of notochord, entoderm with primary intestinal cavity
  • Frog, early tail bud stage, t.s. with neural tube, notochord
  • Frog, early tail bud stage, sagittal l.s. with primordium of brain, ntestine, segmentation of mesoderm
  • Frog, hatching stage, t.s. through region of head or gills
  • Frog, hatching stage, t.s. through region of midbody
  • Frog, young tadpole, t.s. through head
  • Frog, young tadpole, t.s. through thorax or abdomen
Specifications
Type Frog Embryo
Quantity 10