Learn More
World Precision Instrument Frog Heart Clip, 1.7cm Long, Pkg of 5
SDP
Catalog No. 502521378
Encompass
Frog heart clips, also known as frog heart clamps or holders, is a laboratory surgical instrument used in cardiovascular research involving the hearts small animals. Frog heart clips are primarily used to immobilize and stabilize the frog's heart during procedures. In many cardiovascular experiments, researchers need to remove the heart from a frog while keeping it alive and functional, and this clip secures the heart without damaging it. By immobilizing the heart, they can attach electrodes or sensors to monitor physiological parameters such as heart rate, contractility, or response to different drugs and stimuli or to deliver a controlled stimulation. Frog heart clips are sold in packages of 5, and come in two sizes, 0.6mm diameter loop that 17mm long or 2.5mm diameter loop that is 24mm long. These are surgical grade stainless steel, suitable for autoclaving or chemical sterilization.
Catalog No. 50-252-1378 Supplier World Precision Instrument Supplier No. 501903
Frog heart clips, also known as frog heart clamps or holders, is a laboratory surgical instrument used in cardiovascular research involving the hearts small animals. Frog heart clips are primarily used to immobilize and stabilize the frog's heart during procedures. In many cardiovascular experiments, researchers need to remove the heart from a frog while keeping it alive and functional, and this clip secures the heart without damaging it. By immobilizing the heart, they can attach electrodes or sensors to monitor physiological parameters such as heart rate, contractility, or response to different drugs and stimuli or to deliver a controlled stimulation. Frog heart clips are sold in packages of 5, and come in two sizes, 0.6mm diameter loop that 17mm long or 2.5mm diameter loop that is 24mm long. These are surgical grade stainless steel, suitable for autoclaving or chemical sterilization.