Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ From Field to Flask : Investigating Nitrogen Impact on Waterways
SDPSCI_ED

Catalog No. S001100 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S001100 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001100 Supplier EDVOTEK™ Supplier No. 928
Add to Cart
Add to Cart

Students will explore sources of nitrogen in our environment by testing samples for nitrates and nitrites using the Griess reagent

  • Use simple colorimetric methods to detect and measure nitrate levels in real or simulated water samples.
  • Explore how fertilizer runoff contributes to eutrophication and related health concerns.
  • Interpret nitrate results and propose solutions to reduce nutrient pollution.
  • This project was made possible by a Science Education Partnership Award (SEPA), Grant Number 2R44GM143977-02, from the National Institute of General Medical Sciences (NIGMS) of the National Institutes of Health (NIH).
  • Group Size: For 10 Lab Groups.
  • Kit Includes: Instructions, nitrate and nitrite solutions, Griess indicator solution components, conical tubes, microcentrifuge tubes, and PCR tubes.
  • All You Need: Micropipettes and tips, spectrophotometer, water bath PPE, distilled water, and assorted laboratory glassware. Computer with spreadsheet software highly recommended.
  • Storage: Some components require refrigerator storage.

Specifications

Grade Level High School, Higher Ed
Grade 9+
Class Size 10 Lab Groups
Product Type Environmental Testing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.