Learn More
Med Vet International Front Leg Quick Splints Large Dogs, J0119Z, 20in, 1 Pair
Catalog No. 5000581410
Encompass_Preferred
The latest design in pre-formed splints for small animal use, this splint is made of high impact ABS plastic and is scored for individual sizing. Radiolucent. Excellent replacement for Thomas Schroeder splints. All splints come in a paired left and right leg set. REAR LEG OR TARSAL SPLINT Shaped to follow the hock contour.
Catalog No. 50-005-81410 Supplier Med Vet International Supplier No. JOR119Z
The latest design in pre-formed splints for small animal use, this splint is made of high impact ABS plastic and is scored for individual sizing. Radiolucent. Excellent replacement for Thomas Schroeder splints. All splints come in a paired left and right leg set. REAR LEG OR TARSAL SPLINT Shaped to follow the hock contour.