Learn More
Perkin Elmer US LLC Front Plate for Auto Sipper Accessory for LAMBDA 365
Catalog No. 502105500
Encompass_Preferred
The Front Plate for the Auto Sipper Accessory enables the tube connections between the Auto Sipper and the LAMBDA 365 spectrometer.
Catalog No. 50-210-5500 Supplier Perkin Elmer US LLC Supplier No. N4101026
The Front Plate for the Auto Sipper Accessory enables the tube connections between the Auto Sipper and the LAMBDA 365 spectrometer.