Learn More
Ace Glass, Inc. Bottle, pressure, 950mL, 60psi, complete with #25 front-seal plug
SDP
Supplier:  Ace Glass, Inc. 8648251
Encompass_Preferred
Ace Glass pressure tubes and bottles are the premier glass pressure vessels in the industry. Flat-bottomed bottles are rated at 60psi at room temperature while our tubes are rated at 150psi at as much as 120 degrees C. Due to varying conditions, Ace cannot guarantee glass vessels from breakage under pressure. Observe all laboratory safety procedures including working behind a shield. All capacities are total capacity. Working volumes would be less.
Catalog No. NC1853719