Learn More
DSI LLC Gas Syringe A-2 100ul Luer Lock RN .028"x.006"x2" bevel, open end
Catalog No. 502062377
Encompass_Preferred
Gas Syringe A-2 100ul Luer Lock RN .028"x.006"x2" bevel, open end
Catalog No. 50-206-2377 Supplier DSI LLC Supplier No. 050045LL
Gas Syringe A-2 100ul Luer Lock RN .028"x.006"x2" bevel, open end