Learn More
Lakeside Manufacturing Inc Front Access Waste and Recycling Station, Laminated, (2) Compartments, (2) 23 Gallon Capacity
Catalog No. 502884479
Encompass_Preferred
Waste and recycling stations combine attractive functionality with the durability of stainless steel. The Lakeside 4412 Front Access Waste and Recycling Station is made of laminate, with (2) compartments, and has a (2) 23 gallon capacity. This unit has a durable stainless steel construction for easy-to-clean applications. The cabinets include a stainless steel floor to protect carpeted facilities. Liquid/Cup Refuse units include 7-gal. sealed receptacle to prevent spills. The liquid/Cup Refuse units include a recessed drainage sink to minimize dripping and spillage as waste beverages are poured out. This model has leveling feet that provide easy floor leveling. Laminate models are available in 12 standard finishes and custom finishes. Nobody beats Lakeside’s lead times. Made in the USA.
Catalog No. 50-288-4479 Supplier Lakeside Manufacturing Inc Supplier No. 4412
Add to Cart
Add to Cart
Request a Quote
Waste and recycling stations combine attractive functionality with the durability of stainless steel. The Lakeside 4412 Front Access Waste and Recycling Station is made of laminate, with (2) compartments, and has a (2) 23 gallon capacity. This unit has a durable stainless steel construction for easy-to-clean applications. The cabinets include a stainless steel floor to protect carpeted facilities. Liquid/Cup Refuse units include 7-gal. sealed receptacle to prevent spills. The liquid/Cup Refuse units include a recessed drainage sink to minimize dripping and spillage as waste beverages are poured out. This model has leveling feet that provide easy floor leveling. Laminate models are available in 12 standard finishes and custom finishes. Nobody beats Lakeside’s lead times. Made in the USA.