Learn More
Electron Microscopy Sciences Omniprobe Short-Cut™ Coupons Frontside Thinning, Mo/Cu, 26.5° Angle
SDP
Supplier:  Electron Microscopy Sciences 75976MO
Encompass
TEM sample grid coupon designed specially for use in the Short-Cut™ tool. The center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut.
Material: Mo/Cu
Angle: 26.5°
Catalog No. 50-337-84