Learn More
Electron Microscopy Sciences Frostbite Rapid Coolant Can 10 OZ
SDP
Supplier:  Electron Microscopy Sciences 7259410
Encompass_Preferred
Ozone-safe Frostbite Rapid Coolant contains no CFCs yet maintains the ability to rapidly cool paraffin blocks and tissue. Two quick bursts will ensure deep penetration into both paraffin blocks and fresh tissue specimens. The trigger nozzle and needle tube help to localize spray, reducing excess coverage and eliminating waste. Nonflammable.
Catalog No. 50-192-7085