Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex™ Frosted AC Multi-specimen Collector Dispenser Wall Mount
SDP

Catalog No. 22000779
Change view
Click to view available options
Holds:
Commode Specimen Collectors, Urine Cups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
22-000-779 Commode Specimen Collectors, Urine Cups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-000-779 Supplier Poltex™ Supplier No. SPECDISPFRSTW
Add to Cart
Add to Cart

Holds commode specimen collectors, urine cups and more.

  • Two drawers
  • Frosted acrylic construction
  • Comes with wall mount hardware

Specifications

Product Type Organizer
Includes Screws, Anchors
Width (Metric) 483 mm
Autoclavable Not Autoclavable
For Use With (Application) Urine Collection
Holds Commode Specimen Collectors, Urine Cups
Width (English) 19 in.
Color Clear
Material Acrylic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.