Learn More
Selleck Chemical LLC Frovatriptan Succinate
SDP
Catalog No. 5000003531
Encompass_Preferred
Frovatriptan Succinate(SB 209509 Succinate VML 251 Succinate) is the succinate salt form of frovatriptan a synthetic triptan with serotonin (5-HT) receptor agonist activity especially for the 5-HT1B/1D receptors
Catalog No. 50-000-03531 Supplier Selleck Chemical LLC Supplier No. S58481G
Frovatriptan Succinate(SB 209509 Succinate VML 251 Succinate) is the succinate salt form of frovatriptan a synthetic triptan with serotonin (5-HT) receptor agonist activity especially for the 5-HT1B/1D receptors