Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Frovatriptan succinate hydrate | 158930-17-7 | MFCD18251278 | 99.9% | 379.41 g/mol | C18H25N3O6 | 10 MG
SDP

Catalog No. 5000297816
Encompass_Preferred

Frovatriptan succinate hydrate is the succinate salt hydrate form of frovatriptan, a selective 5-HT1B/1D receptor agonist used in pharmacological and preclinical research. Supplied in small packaged quantities for laboratory use, it is intended for receptor binding and migraine pharmacology studies.

  • High purity, suitable for research applications.
  • White to off-white solid form, easy to handle.
  • CAS number 158930-17-7 for unambiguous identification.
  • Molecular weight 379.41 g/mol.
  • Store sealed at -20°C and protect from moisture.

Catalog No. 50-002-97816 Supplier Medchemexpress LLC Supplier No. HYB1658A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Frovatriptan succinate hydrate is the succinate salt hydrate form of frovatriptan, a selective 5-HT1B/1D receptor agonist used in pharmacological and preclinical research. Supplied in small packaged quantities for laboratory use, it is intended for receptor binding and migraine pharmacology studies.

  • High purity, suitable for research applications.
  • White to off-white solid form, easy to handle.
  • CAS number 158930-17-7 for unambiguous identification.
  • Molecular weight 379.41 g/mol.
  • Store sealed at -20°C and protect from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.