Learn More
TARGETMOL CHEMICALS INC Frovatriptan succinate hydrate
SDP
Catalog No. 502880785
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Frovatriptan succinate hydrate (Frova) is effective in treating the full spectrum of migraine including the associated symptoms of nausea, vomiting, photophobia, and phonophobia. Frovatriptan succinate hydrate can also be used as in mini-prophylaxis in menstrual migraine.Frovatriptan succinate hydrate is a potent, high affinity, selective and orally active 5-HT1B, HT1D receptor agonist and a moderately potent 5-HT7 receptor agonist, with pKi values of 8.6, 8.4, and 6.7, respectively. Purity 99.95%
Catalog No. 50-288-0785 Supplier TARGETMOL CHEMICALS INC Supplier No. T1132625MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Frovatriptan succinate hydrate (Frova) is effective in treating the full spectrum of migraine including the associated symptoms of nausea, vomiting, photophobia, and phonophobia. Frovatriptan succinate hydrate can also be used as in mini-prophylaxis in menstrual migraine.Frovatriptan succinate hydrate is a potent, high affinity, selective and orally active 5-HT1B, HT1D receptor agonist and a moderately potent 5-HT7 receptor agonist, with pKi values of 8.6, 8.4, and 6.7, respectively. Purity 99.95%