Learn More
Lonza Walkersville NHAC-kn – Human Knee Articular Chondrocytes
Supplier:  Lonza Walkersville CC2550
Encompass
Clonetics Normal Human Articular Chondrocytes (NHAC-kn) are isolated from tissue taken from the human knee. Cryopreserved Clonetics Normal Human Articular Chondrocytes are guaranteed through 15 population doublings. After differentiation, they are guaranteed positive for collagen type II and sulfated proteoglycans. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased. Applications:OsteoarthritisCartilage failureCellular matrix linkage ruptureChondrocyte protein synthesis inhibition Chondrocyte apoptosisContent:1 x Cryopreserved ampule of NHAC-kn containing ≥750,000 cells
Catalog No. NC9930469
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.