Learn More
Lascar Electronics Inc Pack of 10 Frozen Goods Temp & RH Logger
SDP
Catalog No. NC1856687
Encompass_Preferred
Available in packs of 10, the ultra lost cost single-use EL-CC-2-002 PK10 Frozen Goods data logger measures and stores both temperature and humidity information. The unit comes pre-configured with High -16°C and Low -20°C Alarms - simply press the button to start logging. During a logging session, pressing the button will light up the status LEDs to give a quick indication of whether the temperature has exceeded an alarm condition at any point. Once your session is completed, tear open the IP67-rated plastic wallet to reveal the USB connector and plug it in to your PC. Use the free EasyLog CC software for Windows to download and graph the data.Please Note Whilst in this version of the logger temperature alarms are fixed, and humidity alarms are not set, Lascar can also provide custom configurations of this logger where both temperature and humidity alarms are set to your specifications. Please contact our Sales Team if this is of interest.
Catalog No. NC1856687 Supplier Lascar Electronics Inc Supplier No. ELCC2002 PK10
Available in packs of 10, the ultra lost cost single-use EL-CC-2-002 PK10 Frozen Goods data logger measures and stores both temperature and humidity information. The unit comes pre-configured with High -16°C and Low -20°C Alarms - simply press the button to start logging. During a logging session, pressing the button will light up the status LEDs to give a quick indication of whether the temperature has exceeded an alarm condition at any point. Once your session is completed, tear open the IP67-rated plastic wallet to reveal the USB connector and plug it in to your PC. Use the free EasyLog CC software for Windows to download and graph the data.Please Note Whilst in this version of the logger temperature alarms are fixed, and humidity alarms are not set, Lascar can also provide custom configurations of this logger where both temperature and humidity alarms are set to your specifications. Please contact our Sales Team if this is of interest.